Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

329517

Sigma-Aldrich

2,6,6-Trimethyl-2-cyclohexene-1,4-dione

98%

Synonym(s):

4-Oxoisophorone, Ketoisophorone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O2
CAS Number:
Molecular Weight:
152.19
Beilstein:
2207030
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.491 (lit.)

bp

222 °C (lit.)
92-94 °C/11 mmHg (lit.)

mp

26-28 °C (lit.)

functional group

ketone

SMILES string

CC1=CC(=O)CC(C)(C)C1=O

InChI

1S/C9H12O2/c1-6-4-7(10)5-9(2,3)8(6)11/h4H,5H2,1-3H3

InChI key

AYJXHIDNNLJQDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6,6-Trimethyl-2-cyclohexene-1,4-dione is also known as 4-ketoisophorone and is the major component of saffron spice. It is a cyclic olefin and was reported as a product of the thermal degradation of β-carotene in aqueous suspension.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Description of volatile compounds generated by the degradation of carotenoids in paprika, tomato and marigold oleoresins.
Rios JJ, et al.
Food Chemistry, 106(3), 1145-1153 (2008)
Worldwide market screening of saffron volatile composition.
Maggi L, et al.
Journal of the Science of Food and Agriculture, 89(11), 1950-1954 (2011)
Mohamed-Elamir F Hegazy et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 63(5-6), 403-408 (2008-08-02)
Stereospecific olefin (C=C) and carbonyl (C=O) reduction of the readily available prochiral compound ketoisophorone (2,2,6-trimethyl-2-cyclohexene-1,4-dione) (1) by Marchantia polymorpha and Nicotiana tabacum cell suspension cultures produce the chiral products (6R)-levodione (2), (4R,5S)-4-hydroxy-3,3,5-trimethylcyclohexanone (3), and (4R,6R)-actinol (4) as well as the
Stefano Raimondi et al.
Journal of biotechnology, 156(4), 279-285 (2011-09-22)
Old yellow enzymes (OYEs, EC 1.6.99.1) are flavin-dependent oxidoreductases that catalyze the stereoselective trans-hydrogenation of the double bond, representing a promising alternative to metal-based catalysis. Bioconversion of ketoisophorone (KIP) by 28 non-conventional yeasts belonging to 16 different species was investigated.
Marta Goretti et al.
Bioresource technology, 102(5), 3993-3998 (2011-01-15)
The bioreduction of α,β-unsaturated ketones (ketoisophorone, 2-methyl- and 3-methyl-cyclopentenone) and aldehydes [(S)-(-)-perillaldehyde and α-methyl-cinnamaldehyde] by 23 "non-conventional" yeasts (NCYs) belonging to 21 species of the genera Candida, Cryptococcus, Debaryomyces, Hanseniaspora, Kazachstania, Kluyveromyces, Lindnera, Nakaseomyces, Vanderwaltozyma, and Wickerhamomyces was reported. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service