Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL89328

Isomenthone

phyproof® Reference Substance

Synonym(s):

cis-2-Isopropyl-5-methylcyclohexanone, cis-p-Menthan 3-one, cis-Menthone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (GC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O=C1C(CCC(C1)C)C(C)C

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3

InChI key

NFLGAXVYCFJBMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Essential Oil Biosynthesis: Research on the accumulation and biosynthesis of volatile oils, including Isomenthone, in Agastache rugosa, provides insights into the metabolic pathways and enzyme activities involved, essential for biotechnological applications and drug design (Shu et al., 2024).

  • Optimizing Essential Oil Yield: A study evaluating the effects of harvest season and cultivation method on the yield and composition of essential oils from Clinopodium nepeta subsp. spruneri, emphasizes the role of Isomenthone in achieving optimal bioactivity and quality for pharmaceutical uses (Vlachou et al., 2023).

  • Comprehensive Oil Quality Assessment: Detailed analysis of peppermint oils and their commercial products highlights the importance of Isomenthone in ensuring the comprehensive quality and specificity of bioactive compounds, facilitating advanced pharmaceutical research and development (Wang et al., 2024).

  • Impact of Extraction Conditions: Investigation into the effects of hydrodistillation time on the yield, composition, and antioxidant activity of mint essential oil reveals Isomenthone′s significant impact on the therapeutic qualities and efficiency of natural extracts used in biochemistry and pharmaceutical formulations (Marques et al., 2023).

Other Notes

This compound is commonly found in plants of the genus: mentha

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service