Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

329347

Sigma-Aldrich

Ethanol-2,2,2-d3

99 atom % D

Synonym(s):

Ethyl alcohol-2,2,2-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH2OH
CAS Number:
Molecular Weight:
49.09
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.3611 (lit.)

bp

78 °C (lit.)

density

0.835 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3

InChI key

LFQSCWFLJHTTHZ-FIBGUPNXSA-N

General description

Ethanol-2,2,2-d3 is an isotope-labeled analog of ethanol, wherein methyl protons are replaced by deuterium.

Application

Ethanol-2,2,2-d3 can be used as a deuterated solvent in NMR-based research and analysis.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The orientations and dynamics of ethanol guest molecules in the clathrate of Dianin's compound: A 2H-nuclear magnetic resonance single crystal study
Bernhard T, et al.
Molecular Physics, 77, 1123-1152 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service