Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328693

ALNOCHROMIX

box of 10 packs (10×3.1 oz PACKS)

Synonym(s):

Godax Nochromix, Metal-free cleaner, glass cleaner, non-carcinogenic glass cleaner, persulfate based oxidizing addative, sufactant-free detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

packaging

box of 10 packs (10×3.1 oz PACKS)

manufacturer/tradename

Alconox, Inc. 2510 (Previously Godax 19-010)

technique(s)

protein quantification: suitable

General description

Concentrated inorganic persulfate based additive to sulfuric acid to make a metal-free and surfactant-free cleaner. Excellent for cleaning glassware used in tissue culture, trace metal, and enzyme kinetics research; as well as other sensitive glass cleaning applications.

Directions for use : Pour 1-2 pouches of ALNOCHROMIX crystals into 98% concentrated sulfuric acid container (2.5L or 1gal capacity).

Features and Benefits

  • ALNOCHROMIX™ is an inorganic persulfate based oxidizing acid additive for cleaning glassware.
  • Metal-free and surfactant-free cleaning agent that removes difficult organic, cyanide, and metal residues and odors.
  • Nonmetallic and non-carcinogenic - ideal for sensitive glass cleaning applications.

Legal Information

ALNOCHROMIX is a trademark of Alconox, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcos Francisco Núñez et al.
Molecular biology of the cell, 31(7), 667-682 (2019-12-27)
B-cells become activated by ligands with varying valency and mode of presentation to the B-cell receptor (BCR). We previously demonstrated that clustering the immunoglobulin M (IgM) isotype of BCR with an artificial soluble cross-linker stabilized an ordered phase-like domain that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service