Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

325333

Sigma-Aldrich

3-Chloro-2-hydroxy-1-propanesulfonic acid sodium salt hydrate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(OH)CH2SO3Na · xH2O
CAS Number:
Molecular Weight:
196.59 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

256 °C (dec.) (lit.)

functional group

chloro
sulfonic acid

SMILES string

O.[Na+].OC(CCl)CS([O-])(=O)=O

InChI

1S/C3H7ClO4S.Na.H2O/c4-1-3(5)2-9(6,7)8;;/h3,5H,1-2H2,(H,6,7,8);;1H2/q;+1;/p-1

InChI key

ZPFGAXXLEFTBEU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

3-Chloro-2-hydroxy-1-propanesulfonic acid sodium salt hydrate is a novel anionic agent.

Application

3-Chloro-2-hydroxy-1-propanesulfonic acid sodium salt hydrate was used:
  • to prepare negatively charged probe particles to investigate effect of nanoporosity of cellulosic fibers on their streaming potential and their interactions with cationic polyelectrolytes
  • to prepare surface-derivatized microcrystalline cellulose (MCC) particles having either a strong positive or a strong negative zeta potential

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charge and the dry-strength performance of polyampholytes: Part 2. Colloidal effects.
Hubbe MA, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 301(1), 23-32 (2007)
Consequences of the nanoporosity of cellulosic fibers on their streaming potential and their interactions with cationic polyelectrolytes.
Hubbe MA, et al.
Cellulose, 14(6), 655-671 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service