Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

317713

Sigma-Aldrich

Allyl phenyl sulfone

98%

Synonym(s):

(2-Propen-1-ylsulfonyl)benzene, (2-Propenylsulfonyl)benzene, (Allylsulfonyl)benzene, Phenyl 2-propenyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2CH=CH2
CAS Number:
Molecular Weight:
182.24
Beilstein:
1863561
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.548 (lit.)

bp

110-113 °C/0.5 mmHg (lit.)

density

1.189 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCS(=O)(=O)c1ccccc1

InChI

1S/C9H10O2S/c1-2-8-12(10,11)9-6-4-3-5-7-9/h2-7H,1,8H2

InChI key

KYPIULIVYSQNNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T M Ko et al.
Biomaterials, 14(9), 657-664 (1993-07-01)
Canine platelet adhesion on sulphur dioxide and allyl phenyl sulphone plasma-treated low density polyethylene (LDPE) was studied. Both 5 W plasma excitation energy-treated surfaces showed the highest S:C atomic ratio by ESCA analysis. Surface hydrophilicity increased drastically after plasma treatment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service