Skip to Content
MilliporeSigma
Allyl phenyl sulfone purum, ≥98.0% (GC)

Allyl phenyl sulfone

Allyl phenyl sulfone purum, ≥98.0% (GC)
Linear Formula:
C6H5SO2CH2CH=CH2
CAS Number:
Molecular Weight:
182.24
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing