Skip to Content
MilliporeSigma
All Photos(1)

Documents

308323

Sigma-Aldrich

cis-Stilbene oxide

97%

Synonym(s):

cis-2,3-Diphenyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O
CAS Number:
Molecular Weight:
196.24
Beilstein:
82737
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

mp

38-40 °C (lit.)

storage temp.

2-8°C

SMILES string

O1[C@H]([C@H]1c2ccccc2)c3ccccc3

InChI

1S/C14H12O/c1-3-7-11(8-4-1)13-14(15-13)12-9-5-2-6-10-12/h1-10,13-14H/t13-,14+

InChI key

ARCJQKUWGAZPFX-OKILXGFUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hesam Arabnejad et al.
Chembiochem : a European journal of chemical biology, 21(13), 1893-1904 (2020-01-22)
The use of enzymes in preparative biocatalysis often requires tailoring enzyme selectivity by protein engineering. Herein we explore the use of computational library design and molecular dynamics simulations to create variants of limonene epoxide hydrolase that produce enantiomeric diols from
Li-Ming Yang et al.
Bioorganic & medicinal chemistry letters, 12(7), 1013-1015 (2002-03-23)
A new series of trans-stilbene benzenesulfonamide derivatives were designed and synthesized as potential antitumor agents. These new compounds were evaluated in the National Cancer Institute's 60 human tumor cell line in vitro screen. Compounds 9-13 were cytotoxic against several cell
Neil Everall et al.
Applied spectroscopy, 64(1), 52-60 (2010-02-06)
Picosecond time-resolved transmission Raman data were acquired for 1 mm thick powder samples of trans-stilbene, and a Monte Carlo model was developed that can successfully model the laser and Raman pulse profiles. Photon migration broadened the incident (approximately 1 ps)
V J Mayani et al.
Chirality, 21(2), 255-261 (2008-06-19)
Chromatographic behavior of nonracemic mixtures, viz., mandelic acid and stilbene oxide as analytes has been studied in detailed by enantiomer self-disproportionation on achiral ordered mesoporous material M41S and regular silica gel as stationary phases. Enantiomer self-disproportionation gave enhanced separation of
Larry Miller et al.
Journal of chromatography. A, 1250, 256-263 (2012-06-26)
Preparative supercritical fluid chromatography (SFC) has become the preferred method for the rapid purification of drug candidates during the pharmaceutical discovery process. This paper will discuss the evaluation of injection techniques for preparative SFC. A thorough evaluation of mixed stream

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service