Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

299111

Sigma-Aldrich

Sodium acetate-13C2,d3

99 atom % D, 99 atom % 13C

Synonym(s):

13C and D Labeled sodium acetate, Acetic-13C2,2,2,2-d3 acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CD313CO2Na
CAS Number:
Molecular Weight:
87.04
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C
99 atom % D

form

solid

technique(s)

bio NMR: suitable

mp

>300 °C (dec.) (lit.)

mass shift

M+5

SMILES string

[Na+].[2H][13C]([2H])([2H])[13C]([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1+1D3,2+1;

InChI key

VMHLLURERBWHNL-SZRYOQPCSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>482.0 °F - closed cup

Flash Point(C)

> 250.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service