Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

493007

Sigma-Aldrich

Methane-13C,d4

99 atom % D, 99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CD4
CAS Number:
Molecular Weight:
21.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

vapor density

0.55 (vs air)

isotopic purity

99 atom % 13C
99 atom % D

bp

−161 °C (lit.)

mp

−183 °C (lit.)

mass shift

M+5

SMILES string

[2H][13C]([2H])([2H])[2H]

InChI

1S/CH4/h1H4/i1+1D4

InChI key

VNWKTOKETHGBQD-PPVWBSHVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Brass hose adapter Z14,681-1 or brass body mini gas regulator Z51,353-9 is recommended.

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-306.4 °F - closed cup

Flash Point(C)

-188 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service