Skip to Content
MilliporeSigma
All Photos(1)

Documents

293814

Sigma-Aldrich

2,3-Diethyl-5-methylpyrazine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N2
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.498 (lit.)

density

0.949 g/mL at 25 °C (lit.)

SMILES string

CCc1ncc(C)nc1CC

InChI

1S/C9H14N2/c1-4-8-9(5-2)11-7(3)6-10-8/h6H,4-5H2,1-3H3

InChI key

PSINWXIDJYEXLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Diethyl-5-methylpyrazine has been identified as a potent odorant in coffee sample and has been evaluated by gas chromatography-olfactometry of decreasing headspace samples.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of roasted coffee powders and brews by gas chromatography-olfactometry of headspace samples.
Semmelroch, Peter, and Werner Grosch.
LWT--Food Science and Technology, 28(3), 310-313 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service