Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

290130

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3CHO
CAS Number:
Molecular Weight:
242.12
Beilstein:
1884058
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.422 (lit.)

bp

37 °C/1.3 mmHg (lit.)

density

1.469 g/mL at 25 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

[H]C(=O)c1cc(cc(c1)C(F)(F)F)C(F)(F)F

InChI

1S/C9H4F6O/c10-8(11,12)6-1-5(4-16)2-7(3-6)9(13,14)15/h1-4H

InChI key

LDWLIXZSDPXYDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Bis(trifluoromethyl)benzaldehyde was used in the synthesis of of a series of meso-3,5-bis(trifluoromethyl)phenyl-substituted expanded porphyrins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soonchul Kang et al.
Chemistry, an Asian journal, 3(12), 2065-2074 (2008-11-14)
Trifluoroacetic acid-catalyzed condensation of pyrrole with electron-deficient and sterically hindered 3,5-bis(trifluoromethyl)benzaldehyde results in the unexpected production of a series of meso-3,5-bis(trifluoromethyl)phenyl-substituted expanded porphyrins including [22]sapphyrin 2, N-fused [22]pentaphyrin 3, [26]hexaphyrin 4, and intact [32]heptaphyrin 5 together with the conventional 5,10,15,20-tetrakis(3,5-bis(trifluoromethyl)phenyl)porphyrin
M M V Ramana et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 152, 165-171 (2015-07-25)
In the present work, isopropyl-6-amino-4-(3,5-bis(trifluoromethyl)phenyl)-5-cyano-2-methyl-4H-pyran-3-carboxylate (4H-pyran analog) has been synthesized by a three component reaction catalyzed by CsOH/γ-Al2O3 and characterized. The interaction of 4H-pyran analog with herring sperm DNA (hs DNA) under physiological conditions (phosphate buffer of pH 7.2) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service