Skip to Content
MilliporeSigma
All Photos(1)

Documents

289612

Sigma-Aldrich

N-Ethylcyclohexylamine

98%

Synonym(s):

N-Cyclohexylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NHC2H5
CAS Number:
Molecular Weight:
127.23
Beilstein:
2070280
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.4 (vs air)

Quality Level

vapor pressure

2 mmHg ( 20 °C)

Assay

98%

autoignition temp.

473 °F

expl. lim.

7.6 %

refractive index

n20/D 1.452 (lit.)

bp

165 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CCNC1CCCCC1

InChI

1S/C8H17N/c1-2-9-8-6-4-3-5-7-8/h8-9H,2-7H2,1H3

InChI key

AGVKXDPPPSLISR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Ethylcyclohexylamine was used in the synthesis of a series of isatin-N-Mannich bases.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of isatin-N-mannich bases.
Varma RS and Nobles WL.
Journal of Heterocyclic Chemistry, 3(4), 462-465 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service