Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

289310

Sigma-Aldrich

Rubidium carbonate

99% (trace metals analysis)

Synonym(s):

Carbonic acid dirubidium, Dirubidium carboxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rb2CO3
CAS Number:
Molecular Weight:
230.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

99% (trace metals analysis)

form

powder

SMILES string

[Rb+].[Rb+].[O-]C([O-])=O

InChI

1S/CH2O3.2Rb/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

WPFGFHJALYCVMO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Rubidium carbonate can be used:
  • As a strong n-dopant to fabricate quantum-dot light-emitting diodes. The addition of rubidium carbonate enhances the thermal stability of Mg-doped ZnO electron transport layer.
  • As a precursor to prepare rubidium-promoted iron catalyst for Fischer–Tropsch synthesis.
  • As a strong base for the cross-coupling reaction between organobismuthines and aryl or heteroaryl halides to synthesize triarylbismuthines.
  • To prepare rubidium borosilicate glasses.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Highly Functionalized Triarylbismuthines by Functional Group Manipulation and Use in Palladium- and Copper-Catalyzed Arylation Reactions
Martin Hebert, et al.
The Journal of Organic Chemistry, 81, 5401-5416 (2016)
Thermal study of melting, transition and crystallization of rubidium and cesium borosilicate glasses
V.E. Eremyashev, et al.
Ceramics International, 42, 18368-18372 (2016)
Fischer-Tropsch synthesis: Characterization Rb promoted iron catalyst
Amitava Sarkar, et al.
Catalysis Letters, 121, 1-11 (2008)
Remarkable lifetime improvement of quantum-dot light-emitting diodes by incorporating rubidium carbonate in metal-oxide electron transport layers
Yujin Lee, et al.
Journal of Materials Chemistry, 7, 10082-10091 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service