Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43858

Sigma-Aldrich

Rubidium carbonate

for synthesis

Synonym(s):

Rubidium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CO3Rb2
CAS Number:
Molecular Weight:
230.94
MDL number:
UNSPSC Code:
12352302
EC Index Number:
209-530-9
NACRES:
NA.22
grade:
for synthesis
form:
powder
solubility:
4500 g/L

grade

for synthesis

Quality Level

form

powder

potency

2625 mg/kg LD50, oral (Rat)

pH

11.7 (20 °C, 50 g/L in H2O)

mp

837 °C

solubility

4500 g/L

storage temp.

2-30°C

SMILES string

[Rb+].[Rb+].[O-]C(=O)[O-]

InChI

1S/CH2O3.2Rb/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

WPFGFHJALYCVMO-UHFFFAOYSA-L

Analysis Note

Assay: ≥ 98.0 %
Identity (ICP-OES): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service