Skip to Content
MilliporeSigma
All Photos(1)

Documents

288810

Sigma-Aldrich

3-Methoxyphenylacetone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2COCH3
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.5252 (lit.)

bp

258-260 °C (lit.)

density

1.911 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(CC(C)=O)c1

InChI

1S/C10H12O2/c1-8(11)6-9-4-3-5-10(7-9)12-2/h3-5,7H,6H2,1-2H3

InChI key

RMMRRRLPDBJBQL-UHFFFAOYSA-N

Application

3-Methoxyphenylacetone was used in the synthesis of 4-amino-3-methoxypropiophenone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

First synthesis of 3-methoxy-4-aminopropiophenone.
J. Chem. Res. (M), 36(9), 543-544 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service