Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

288144

Sigma-Aldrich

Tricarbonyldichlororuthenium(II) dimer

Synonym(s):

CORM-2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ru(CO)3Cl2]2
CAS Number:
Molecular Weight:
512.01
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

core: ruthenium
reagent type: catalyst

SMILES string

[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].Cl[Ru]Cl.Cl[Ru]Cl

InChI

1S/6CO.4ClH.2Ru/c6*1-2;;;;;;/h;;;;;;4*1H;;/q;;;;;;;;;;2*+2/p-4

InChI key

JYHHJVKGDCZCCL-UHFFFAOYSA-J

Application

Carbon monoxide-releasing molecule used for pharamological studies including:
  • Enhancement of coagulation and attenuation of vulnerability to fibrinolysis
  • Investigations of P2X4 as a possible target
  • Possible modification of thrombus growth or disintegration
  • Enhances fibrinogen as a substrate for thrombin
  • Regulation of ion transport by gasotransmitters

Used as a CO donor for reactive oxygen species mediated bacterial killing
Tricarbonyldichlororuthenium(II) dimer can be used in the deuteration reaction for labeling secondary amines. It can also be used for the synthesis of racemization catalysts by reacting with cyclopenta[l]phenanthrenyl and cyclopenta[a]acenaphthylenyl ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step exchange-labelling of piperidines, piperazines and dialkylamines with deuterium oxide: catalysis by various ruthenium complexes.
Alexakis E, et al.
Tetrahedron Letters, 46(25), 4291-4293 (2005)
Cyclopenta [l] phenanthrenyl and Cyclopenta [a] acenaphthylenyl Half-Sandwich Complexes of Ruthenium as Racemization Catalysts for Secondary Alcohols.
Mavrynsky D, et al.
Organometallics, 28(2), 598-605 (2008)
Vance G Nielsen et al.
International journal of molecular sciences, 21(6) (2020-03-22)
Using thrombelastography to gain mechanistic insights, recent investigations have identified enzymes and compounds in Naja and Crotalus species' neurotoxic venoms that are anticoagulant in nature. The neurotoxic venoms of the four extant species of Dendroaspis (the Black and green mambas)
Po-Min Yang et al.
Vascular pharmacology, 87, 209-218 (2016-10-11)
The production of nitric oxide (NO) by endothelial NO synthase (eNOS) plays a major role in maintaining vascular homeostasis. This study elucidated the potential role of carbon monoxide (CO)-releasing molecules (CORMs) in NO production and explored the underlying mechanisms in
Francisco J Ibáñez et al.
Frontiers in microbiology, 8, 2108-2108 (2017-11-23)
Heme oxygenase-1 (HO-1) is an inducible enzyme that is expressed in response to physical and chemical stresses, such as ultraviolet radiation, hyperthermia, hypoxia, reactive oxygen species (ROS), as well as cytokines, among others. Its activity can be positively modulated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service