Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

683191

Sigma-Aldrich

Chiralyst P280

Umicore, 95%

Synonym(s):

Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer, Dichloro(1,5-cyclooctadiene)ruthenium(II), (1,5-Cyclooctadiene)ruthenium(II) chloride, Ruthenium(II) chloride 1,5-cyclooctadiene complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ru(COD)Cl2]n
CAS Number:
Molecular Weight:
280.16 (as monomer)
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

SMILES string

[Ru+2].[Cl-].[Cl-].C1CC=CCCC=C1

InChI

1S/C8H12.2ClH.Ru/c1-2-4-6-8-7-5-3-1;;;/h1-2,7-8H,3-6H2;2*1H;/q;;;+2/p-2

InChI key

DMRVBCXRFYZCPR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service