Skip to Content
MilliporeSigma
All Photos(2)

Documents

288128

Sigma-Aldrich

cis-Bis(2,2′-bipyridine)dichlororuthenium(II) hydrate

Synonym(s):

cis-Ru(bpy)2Cl2.xH2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16Cl2N4Ru · xH2O
CAS Number:
Molecular Weight:
484.34 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

reaction suitability

reaction type: C-H Activation
reagent type: catalyst

SMILES string

[H]O[H].Cl[Ru]Cl.c1ccc(nc1)-c2ccccn2.c3ccc(nc3)-c4ccccn4

InChI

1S/2C10H8N2.2ClH.H2O.Ru/c2*1-3-7-11-9(5-1)10-6-2-4-8-12-10;;;;/h2*1-8H;2*1H;1H2;/q;;;;;+2/p-2

InChI key

XBOQABVNRXVAKI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

cis-Bis(2,2′-bipyridine)dichlororuthenium(II) hydrate was used in the preparation of [Ru(m-) 2,2′-bipyridine)3]2+(PF6-)2. It was used to prepare an intermediate for the development of nanocomposite junctions with Au nanoparticles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electroluminescence in Ruthenium(II) Complexes
Bernhard S, et al
Journal of the American Chemical Society, 124, 13624-13628 null
Syntheses,Crystal Structures, and Spectral Properties of a Series of 3,8-Bisphenyl-1,10-phenanthroline Derivatives: Precursors of 3,8-Bis(4-mercaptophenyl)-1,10-phenanthroline and Its Ruthenium(II) Complex for Preparing Nanocomposite Junctions with Gold Nanoparticles between 1 ?m Gap Gold Electrodes
Huang W, et al.
Inorganic Chemistry, 47, 468?480-468?480 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service