Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

284459

Sigma-Aldrich

Trimethylsilyl acetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2Si(CH3)3
CAS Number:
Molecular Weight:
132.23
Beilstein:
1744463
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1 (vs air)

Quality Level

vapor pressure

35 mmHg ( 30 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.388 (lit.)

bp

107.5 °C (lit.)

mp

−32 °C (lit.)

density

0.882 g/mL at 25 °C (lit.)

SMILES string

CC(=O)O[Si](C)(C)C

InChI

1S/C5H12O2Si/c1-5(6)7-8(2,3)4/h1-4H3

InChI key

QHUNJMXHQHHWQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel base-initiated reactions of N-substituted pyridinium salts.
Journal of Fluorine Chemistry, 77(2), 161-168 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service