Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

274801

Sigma-Aldrich

3′,4′-(Methylenedioxy)acetophenone

98%

Synonym(s):

1-(1,3-Benzodioxol-5-yl)ethanone, 1-(1,3-Dioxaindan-5-yl)ethan-1-one, 1-(Benz[d][1,3]dioxol-5-yl)ethanone, 1-(Benzo[d][1,3]dioxol-5-yl)ethanone, 1-(Benzodioxol-5-yl)ethanone, 1-Acetyl-3,4-(methylenedioxy)benzene, 3,4-(Methylenedioxy)phenyl methyl ketone, 5-Acetyl-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O3
CAS Number:
Molecular Weight:
164.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

87-89 °C (lit.)

SMILES string

CC(=O)c1ccc2OCOc2c1

InChI

1S/C9H8O3/c1-6(10)7-2-3-8-9(4-7)12-5-11-8/h2-4H,5H2,1H3

InChI key

BMHMKWXYXFBWMI-UHFFFAOYSA-N

Application

3′,4′-(Methylenedioxy)acetophenone was used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Marson et al.
Journal of analytical toxicology, 24(1), 17-21 (2000-02-02)
The present paper describes investigations following the analysis of a urine specimen containing important amounts of an unknown substance detected by gas chromatography-mass spectrometry (GC-MS) analysis. FPIA analysis was positive (cutoff 0.3 mg/L) and Triage 8 rapid test was negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service