Skip to Content
MilliporeSigma
All Photos(2)

Documents

396583

Sigma-Aldrich

4′-(Methylthio)acetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SC6H4COCH3
CAS Number:
Molecular Weight:
166.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

80-82 °C (lit.)

SMILES string

CSc1ccc(cc1)C(C)=O

InChI

1S/C9H10OS/c1-7(10)8-3-5-9(11-2)6-4-8/h3-6H,1-2H3

InChI key

JECUZQLBQKNEMW-UHFFFAOYSA-N

General description

4′-(Methylthio)acetophenone (4-(Methylthio)acetophenone, p-(methylthio) acetophenone, 4-MTAP) is a sulphur containing organic building block. It is a key intermediate in drug synthesis. Its industrial preparation, via Friedel-Crafts acylation reaction in the presence of aluminium chloride as catalyst and acetyl chloride as acylating agent has been reported. It has been prepared by the acetylation reaction of thioanisole with acetic anhydride in the presence of solid acid catalyst.

Application

4′-(Methylthio)acetophenone may be used for the synthesis of Vioxx (Rofecoxib), an non-steroidal anti-inflammatory drug (NSAID). It may be used in the synthesis of (E)-1-[4-(methylsulfanyl)phenyl]-3-phenylprop-2-en-1-one.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(E)-1-[4-(Methylsulfanyl) phenyl]-3-phenylprop-2-en-1-one.
Thiruvalluvar A, et al.
Acta Crystallographica Section E, Structure Reports Online, 64(7), o1263-o1263 (3008)
Experimental and theoretical analysis of Friedel-Crafts acylation of thioanisole to 4-(methylthio) acetophenone using solid acids.
Yadav GD and Bhagat RD.
J. Mol. Catal. A: Chem., 235(1), 98-107 (2005)
Liquid phase acetylation of thioanisole with acetic anhydride to 4-(methylthio) acetophenone (4-MTAP) using H-beta catalyst.
Sawant DP and Halligudi SB.
Catalysis Communications, 5(11), 659-663 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service