Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

280577

Sigma-Aldrich

cis-13-Docosenoamide

Synonym(s):

Erucamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)11CONH2
CAS Number:
Molecular Weight:
337.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

beads

composition

erucamide, >85%

mp

79-81 °C (lit.)

functional group

amide

storage temp.

−20°C

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCCCCC(N)=O

InChI

1S/C22H43NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h9-10H,2-8,11-21H2,1H3,(H2,23,24)/b10-9-

InChI key

UAUDZVJPLUQNMU-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

cis-13-Docosenoamide (Erucamide) was employed as:
  • standard in determination of fatty acid amides in polyethylene packaging film by GC/MS
  • slip agent for polymers to reduce their friction coefficient and to make films easier to handle

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Wakamatsu et al.
Biochemical and biophysical research communications, 168(2), 423-429 (1990-04-30)
Erucamide (13-docosenamide) was found to be the major bovine mesentery angiogenic lipid as assessed by chorioallantoic membrane (CAM) assay. Two micrograms of this lipid caused angiogenesis in the assay. Angiogenic activity of this naturally occurring lipid was also found by
Anders Hamberger et al.
Neurochemical research, 28(2), 177-185 (2003-03-01)
The aim of this study was to isolate a compound from blood plasma that inhibits intestinal diarrhea and that appears also to regulate fluid volumes in other organs. The isolation procedure included lipid extraction, liquid chromatography, and gas chromatography. The
Robert N Tuttle et al.
Applied and environmental microbiology, 85(8) (2019-02-10)
Thousands of natural products have been identified from cultured microorganisms, yet evidence of their production in the environment has proven elusive. Technological advances in mass spectrometry, combined with public databases, now make it possible to address this disparity by detecting
Jianjun Wang et al.
Scientific reports, 7(1), 14667-14667 (2017-11-09)
A series of alkyl amides was detected and identified in the sedimentary record from an archaeological site at Yuchisi, Mengcheng, Anhui, China. The alkyl amides profiles change abruptly at the depth corresponding to the transition between two prehistoric cultures, which
J O Chleboun et al.
Biochemical and biophysical research communications, 185(2), 510-516 (1992-06-15)
An adequate collateral circulation is crucial to tissue survival subsequent to proximal major arterial occlusion. The precise mechanism of collateral blood vessel development and the biochemical mediators involved in this process are unknown. To evaluate the influence of a number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service