270369
1,1,2-Trichlorotrifluoroethane
HPLC grade, ≥99.9%
Synonym(s):
1,1,2-Trichloro-1,2,2-trifluoroethane, 1,1,2-Trichloro-trifluoroethane, Genetron® 113
About This Item
Recommended Products
grade
HPLC grade
vapor density
6.5 (vs air)
vapor pressure
5.5 psi ( 20 °C)
Assay
≥99.9%
form
liquid
autoignition temp.
1256 °F
technique(s)
HPLC: suitable
impurities
<0.010% water
evapn. residue
<0.0005%
refractive index
n20/D 1.358 (lit.)
bp
47-48 °C (lit.)
mp
−35 °C (lit.)
density
1.57 g/mL at 25 °C (lit.)
λ
H2O reference
UV absorption
λ: 231 nm Amax: 1.00
λ: 240 nm Amax: 0.20
λ: 260-400 nm Amax: 0.01
SMILES string
FC(F)(Cl)C(F)(Cl)Cl
InChI
1S/C2Cl3F3/c3-1(4,6)2(5,7)8
InChI key
AJDIZQLSFPQPEY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Ozone 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
383.0 °F
Flash Point(C)
195 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service