Skip to Content
MilliporeSigma
All Photos(1)

Documents

613894

Sigma-Aldrich

Dichlorofluoromethane-d

98 atom % D

Synonym(s):

Freon® 21-d

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDFCl2
Molecular Weight:
103.93
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

form

gas

mass shift

M+1

SMILES string

[2H]C(F)(Cl)Cl

InChI

1S/CHCl2F/c2-1(3)4/h1H/i1D

InChI key

UMNKXPULIDJLSU-MICDWDOJSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Freon is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Gas cylinderExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Ozone 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service