Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

263419

Sigma-Aldrich

3,5-Bis(trifluoromethyl)phenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
272.14
Beilstein:
6813447
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

121-123 °C (lit.)

solubility

methanol: soluble 25 mg/mL, clear, colorless

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)Cc1cc(cc(c1)C(F)(F)F)C(F)(F)F

InChI

1S/C10H6F6O2/c11-9(12,13)6-1-5(3-8(17)18)2-7(4-6)10(14,15)16/h1-2,4H,3H2,(H,17,18)

InChI key

PAWSKKHEEYTXSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Bis(trifluoromethyl)phenylacetic acid has been used:
  • as building block to synthesize the pentaamine and bis-heterocyclic libraries
  • in determination of ionic perfluorinated substances and telomers in leachates from landfills and sediment samples

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adel Nefzi et al.
Bioorganic & medicinal chemistry letters, 19(17), 5169-5175 (2009-07-28)
Combinatorial chemistry offers a unique opportunity for the synthesis and screening of large numbers of compounds and significantly enhances the prospect of finding new drugs. Collaborative efforts with the Tuberculosis Antimicrobial Acquisition & Coordinating Facility (TAACF), have led to the
Trine Eggen et al.
The Science of the total environment, 408(21), 5147-5157 (2010-08-11)
Landfills have historically remained the most common methods of organized waste disposal and still remain so in many regions of the world. Thus, they may contain wastes resulting from several decades of disposal and decomposition with subsequent release of organic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service