Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

262579

Sigma-Aldrich

2-(Bromomethyl)tetrahydro-2H-pyran

98%

Synonym(s):

Tetrahydro-2H-pyran-2-methyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BrO
CAS Number:
Molecular Weight:
179.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.489 (lit.)

bp

153 °C (lit.)

mp

−59-−58 °C (lit.)

density

1.397 g/mL at 25 °C (lit.)

SMILES string

BrCC1CCCCO1

InChI

1S/C6H11BrO/c7-5-6-3-1-2-4-8-6/h6H,1-5H2

InChI key

MHNWCBOXPOLLIB-UHFFFAOYSA-N

General description

Cross-coupling reaction of 2-(bromomethyl)tetrahydro-2H-pyran with potassium heteroaryltrifluoroborates has been investigated.

Application

2-(Bromomethyl)tetrahydro-2H-pyran has been used in the preparation of tellurated heterocycles, 2-[(2-thienyltelluro)methyl]tetrahydrofuran and [(2-thienyltelluro)methyl]tetrahydro-2H-pyran.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tellurated heterocycles, 2-[(2-thienyltelluro)methyl]tetrahydrofuran and [(2-thienyltelluro)methyl]tetrahydro-2H-pyran: Synthesis and complexation reactions with Pd(II), Pt(II), Hg(II), Ru(II) and Cu(I)
Bali S, et al.
Journal of Organometallic Chemistry, 691(18), 3788-3796 (2006)
Gary A Molander et al.
Organic letters, 12(24), 5783-5785 (2010-11-26)
A method for the cross-coupling of alkyl electrophiles with various potassium aryl- and heteroaryltrifluoroborates has been developed. Nearly stoichiometric amounts of organoboron species could be employed to cross-couple a large variety of challenging heteroaryl nucleophiles. Several functional groups were tolerated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service