Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

252069

Sigma-Aldrich

Chlorodifluoroacetic anhydride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCF2CO)2O
CAS Number:
Molecular Weight:
242.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.347 (lit.)

bp

96-97 °C (lit.)

density

1.395 g/mL at 25 °C (lit.)

functional group

anhydride
chloro
ester
fluoro

SMILES string

FC(F)(Cl)C(=O)OC(=O)C(F)(F)Cl

InChI

1S/C4Cl2F4O3/c5-3(7,8)1(11)13-2(12)4(6,9)10

InChI key

VBJIFLOSOQGDRZ-UHFFFAOYSA-N

General description

Photolysis of the vapour of chlorodifluoroacetic anhydride has been studied over the range 19-201°C using 248nm radiation.

Application

Chlorodifluoroacetic anhydride has been used:
  • for dual element acetylation tagging derivatization of amines by using microwave sustained helium emission detector
  • as derivatization reagent in determination of estrone and 17β-estradiol in hair by GC-MS

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Photochemistry of anhydrides. Part 6.-Photolysis of chlorodifluoroacetic anhydride,(CF 2 ClCO) 2 O.
Watkins PE and Whittle E.
J. Chem. Soc., Faraday, 80(8), 2323-2329 (1984)
M H Choi et al.
The Analyst, 125(4), 711-714 (2000-07-13)
An efficient procedure is described for the determination of estrone and 17 beta-estradiol in hair by gas chromatography-mass spectrometry (GC-MS). The method involves alkyloxycarbonylation with isobutyl chloroformate (isoBCF) of phenolic hydroxy groups after alkaline digestion of hair samples. The resulting
Multi-element taggant and dual homolog derivatization concepts in GC/MED.
Hagen DF, et al.
Spectrochimica Acta. Part B: Atomic Spectroscopy, 40(1), 335-347 (1985)
Courtney T Jollymore-Hughes et al.
Bioorganic & medicinal chemistry, 24(21), 5270-5279 (2016-09-18)
Association of cholinesterase with β-amyloid plaques and tau neurofibrillary tangles in Alzheimer's disease offers an opportunity to detect disease pathology during life. Achieving this requires development of radiolabelled cholinesterase ligands with high enzyme affinity. Various fluorinated acetophenone derivatives bind to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service