Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300837

Sigma-Aldrich

Methyl chlorodifluoroacetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClF2CCOOCH3
CAS Number:
Molecular Weight:
144.50
Beilstein:
1756072
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.349 (lit.)

bp

79-81 °C (lit.)

density

1.37 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(F)(F)Cl

InChI

1S/C3H3ClF2O2/c1-8-2(7)3(4,5)6/h1H3

InChI key

AWUPLMYXZJKHEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl chlorodifluoroacetate is a trifluoromethylating agent.

Application

Methyl chlorodifluoroacetate was used in preparation of 2,2-difluoro-3-hydroxyesters and gem-difluorocyclopropane.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyl chlorodifluoroacetate a convenient trifluoromethylating agent.
De-Bao S, et al.
Tetrahedron Letters, 32(52), 7689-7690 (1991)
Electrochemical, nickel-catalyzed Reformatsky reaction with methyl chlorodifluoroacetate
Mcharek S, et al.
Journal of Organometallic Chemistry, 401(1), 211-215 (1991)
Preparation of gem-difluorocyclopropanes via decomposition of methyl chlorodifluoroacetate by alkali metal halides.
Wheaton GA and Burton DJ.
Journal of Fluorine Chemistry, 9(1), 25-44 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service