Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

248975

Sigma-Aldrich

α-Methoxyphenylacetic acid

99%

Synonym(s):

(±)-α-Methoxyphenylacetic acid, O-Methyl-DL-mandelic acid, MOPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)COOH
CAS Number:
Molecular Weight:
166.17
Beilstein:
2209197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

165 °C/18 mmHg (lit.)

mp

69-71 °C (lit.)

functional group

carboxylic acid
ether
phenyl

SMILES string

COC(C(O)=O)c1ccccc1

InChI

1S/C9H10O3/c1-12-8(9(10)11)7-5-3-2-4-6-7/h2-6,8H,1H3,(H,10,11)

InChI key

DIWVBIXQCNRCFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Enantiomers of α-methoxyphenylacetic acid has been resolved as amide derivatives on chiral statinary phases by HPLC. α-Methoxyphenylacetic acid is also known as Trost′s chiral acid.

Application

α-Methoxyphenylacetic acid has been used in the enantioresolution of two diastereomeric esters by HPLC.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resolution of a-arylacetic acid enantiomers on two chiral stationary phases.
McDaniel DM and Snider BG.
Journal of Chromatography A, 404, 123-132 (1987)
2-Methoxy-2-(1-naphthyl) propionic acid, a powerful chiral auxiliary for enantioresolution of alcohols and determination of their absolute configurations by the< sup> 1</sup> H NMR anisotropy method.
Harada N, et al.
Tetrahedron Asymmetry, 11(6), 1249-1253 (2000)
Bin Chang et al.
Journal of clinical microbiology, 53(10), 3318-3324 (2015-08-14)
Streptococcus pneumoniae colonizes the nasopharyngeal mucus in healthy people and causes otitis media, pneumonia, bacteremia, and meningitis. In this study, we analyzed an S. pneumoniae strain that caused 7 repeated pneumonia episodes in an 80-month-old patient with cerebral palsy during
Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at
Weidong Liu et al.
Chembiochem : a European journal of chemical biology, 16(6), 924-929 (2015-03-11)
A meso-diaminopimelate dehydrogenase (DAPDH) from Clostridium tetani E88 (CtDAPDH) was found to have low activity toward the D-amino acids other than its native substrate. Site-directed mutagenesis similar to that carried out on the residues mutated by Vedha-Peters et al. resulted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service