Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

242543

Sigma-Aldrich

Propylamine hydrochloride

≥99%

Synonym(s):

1-Aminopropane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
95.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

solid

mp

160-162 °C (lit.)

solubility

water: soluble 100 mg/mL, clear, colorless

functional group

amine

SMILES string

Cl[H].CCCN

InChI

1S/C3H9N.ClH/c1-2-3-4;/h2-4H2,1H3;1H

InChI key

PYNUOAIJIQGACY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propylamine hydrochloride was used to study the liquid chromatographic behavior of potassium-sparing and loop diuretics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F De Croo et al.
Journal of chromatography, 325(2), 395-411 (1985-06-07)
This paper deals with the specific identification of several thiazide, potassium-sparing and loop diuretics. The liquid chromatographic behaviour of these compounds is studied. Different organic modifiers (methanol, acetonitrile and tetrahydrofuran) are compared in terms of selectivity for the thiazide diuretics.
Rui Tao et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(8), 1996-2007 (2014-03-01)
Drug interaction between inhibitors of monoamine oxidase (MAOIs) and selective serotonin (5-hydroxytryptamine, 5-HT) reuptake (SSRIs) induces serotonin syndrome, which is usually mild but occasionally severe in intensity. However, little is known about neural mechanisms responsible for the syndrome induction and
M Figueiredo-González et al.
Food research international (Ottawa, Ont.), 106, 558-567 (2018-03-28)
In this work, phenol-rich extracts from 'Cornicabra' and 'Picual' virgin-olive oils (EVOOs) were examined, for the first time, to establish their capacity to inhibit key enzymes involved in Alzheimer's disease (AD) (acetylcholinesterase (AChE), butyrylcholinesterase (BuChE) and 5-lipoxygenase (LOX)), major depressive
Sarah W Black et al.
Biological psychiatry, 82(9), 623-633 (2016-12-07)
Narcolepsy, a disorder of rapid eye movement (REM) sleep, is characterized by excessive daytime sleepiness and cataplexy, a loss of muscle tone triggered by emotional stimulation. Current narcolepsy pharmacotherapeutics include controlled substances with abuse potential or drugs with undesirable side
Abu Bin Ihsan et al.
International journal of molecular sciences, 20(18) (2019-09-20)
A series of N-substituted poly(Gly-alter-Val) peptides were successfully synthesized for the systematic evaluation of the micellization behavior of alternating peptides. Three-component polymerization employing an aldehyde, a primary ammonium chloride, and potassium isocyanoacetate afforded four alternating peptides in excellent yields. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service