Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

241962

Sigma-Aldrich

2,3-Pentanedione

97%

Synonym(s):

Acetylpropionyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCOCH3
CAS Number:
Molecular Weight:
100.12
Beilstein:
1699638
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.404 (lit.)

bp

110-112 °C (lit.)

mp

−52 °C (lit.)

density

0.957 g/mL at 25 °C (lit.)

functional group

ketone

storage temp.

2-8°C

SMILES string

CCC(=O)C(C)=O

InChI

1S/C5H8O2/c1-3-5(7)4(2)6/h3H2,1-2H3

InChI key

TZMFJUDUGYTVRY-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Pentanedione (acetylpropionyl) is one of the aroma active compounds of cereal coffee brew.

Application

2,3-Pentanedione can be used as a reactant to synthesize:
  • Bisphenol derivatives by acid-catalyzed condensation reaction with phenols.
  • 2-Ethyl-3-methyl-1H-indole by Pd-catalyzed reaction with aniline under reductive conditions.
  • 2-Ethyl-3-methylquinoxaline by condensation reaction with o-phenylenediamine using citric acid as a catalyst.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Sens. 1B - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - open cup

Flash Point(C)

19 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Małgorzata A Majcher et al.
Journal of agricultural and food chemistry, 61(11), 2648-2654 (2013-02-19)
Cereal coffee is a coffee substitute made mainly from roasted cereals such as barley and rye (60-70%), chicory (15-20%), and sugar beets (6-10%). It is perceived by consumers as a healthy, caffeine free, non-irritating beverage suitable for those who cannot
Regioselective synthesis of renewable bisphenols from 2, 3-pentanedione and their application as plasticizers
Schutyser W, et al.
Green Chemistry, 16(4), 1999-2007 (2014)
Qinoxaline II. A practical efficient and rapid synthesis of new quinoxalines catalyzed by citric acid as a trifunctional Bronsted acid at room temperature under green condition
Sajjadifar S, et al.
International Journal of ChemTech Research, 5(1), 422-429 (2013)
[(S)-BINAP] PdBr2-catalyzed direct synthesis of 2, 3-disubstituted indoles via a tandem reaction between arylamines and α-diketones
Cabrera A, et al.
Tetrahedron Letters, 52(50), 6758-6762 (2011)
Paul Schweiger et al.
Applied microbiology and biotechnology, 87(4), 1415-1426 (2010-04-24)
Two cytosolic NADPH-dependent carbonyl reductases from Gluconobacter oxydans 621H, Gox0644 and Gox1615, were heterologously produced in Escherichia coli. The recombinant proteins were purified to homogeneity and characterized. Gox0644 and Gox1615 were dimers with native molecular masses of 66.1 and 74.5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service