Skip to Content
MilliporeSigma

69043

2,3-Pentanedione

analytical standard

Synonym(s):

Acetylpropionyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH2COCOCH3
CAS Number:
Molecular Weight:
100.12
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
209-984-8
Beilstein/REAXYS Number:
1699638
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

impurities

≤1.0% water

refractive index

n20/D 1.401-1.409, n20/D 1.404 (lit.)

bp

110-112 °C (lit.)

mp

−52 °C (lit.)

density

0.957 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)C(C)=O

InChI

1S/C5H8O2/c1-3-5(7)4(2)6/h3H2,1-2H3

InChI key

TZMFJUDUGYTVRY-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
2,3-Pentanedione may be used as a reference standard in the determination of 2,3-pentanedione in beer samples using headspace-gas chromatographic technique (HS-GC).

Disclaimer

air and light sensitive; packaged under argon


Still not finding the right product?

Explore all of our products under 2,3-Pentanedione


signalword

Danger

Hazard Classifications

Flam. Liq. 2 - Skin Sens. 1B - STOT RE 2

Storage Class

3 - Flammable liquids

flash_point_f

66.2 °F - open cup

flash_point_c

19 °C - open cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library