Skip to Content
MilliporeSigma
All Photos(1)

Documents

239488

Sigma-Aldrich

1,4-Bis(3-aminopropyl)piperazine

≥99%

Synonym(s):

1,4-(3-Aminopropyl)piperazine, 1,4-Piperazinebis(propylamine), 1,4-Piperazinedipropylamine, N,N′-Bis(γ-aminopropyl)piperazine, N,N′-Bis(3-aminopropyl)piperazine, [3-[4-(3-Aminopropyl)piperazin-1-yl]propyl]amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24N4
CAS Number:
Molecular Weight:
200.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.9 (vs air)

Assay

≥99%

form

liquid

refractive index

n20/D 1.502 (lit.)

bp

150-152 °C/2 mmHg (lit.)

mp

15 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

NCCCN1CCN(CCCN)CC1

InChI

1S/C10H24N4/c11-3-1-5-13-7-9-14(10-8-13)6-2-4-12/h1-12H2

InChI key

XUSNPFGLKGCWGN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

The thermodynamic parameters for protonation of 1,4-bis(3-aminopropyl)-piperazine (BAPP) were studied in aqueous solution using a potentiometric technique.

Application

1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

325.4 °F - closed cup

Flash Point(C)

163 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed A El-Sherif et al.
Bioinorganic chemistry and applications, 2012, 984291-984291 (2012-12-12)
Thermodynamic parameters for protonation of 1,4-bis(3-aminopropyl)-piperazine (BAPP) and its metal complexation with some divalent metal ions were determined in aqueous solution at constant ionic strength (0.1 M NaNO(3)) using a potentiometric technique. The order of -ΔG(0) and -ΔH(0) was found to
Jian Cheng et al.
Acta biomaterialia, 30, 155-167 (2015-12-02)
Nano-polyplexes from bioreducible cationic polymers have a massive promise for cancer gene therapy. However, the feasibility of cationic polyurethanes for non-viral gene therapy is so far not well studied. In this work, a linear cationic polyurethane containing disulfide bonds, urethane
Romain Gautier et al.
Advanced materials (Deerfield Beach, Fla.), 31(14), e1807383-e1807383 (2019-02-19)
Hybrid metal halides containing perovskite layers have recently shown great potential for applications in solar cells and light-emitting diodes. Such compounds exhibit quantum confinement effects leading to tunable optical and electronic properties. Thus, broadband white-light emission has been observed from
Chi-Jung Chang et al.
Polymers, 12(9) (2020-09-17)
It is crucial to remove heavy metals and dyes before discharging industrial effluents. Gauze substrate was surface-modified by coating with a polymeric adsorbent and a spray coating of BiOBr photocatalyst to develop a novel dual-functional membrane, polymer/BiOBr-modified gauze, for water
Yanyan Song et al.
Molecular pharmaceutics, 11(7), 2250-2261 (2014-06-04)
A folate-decorated, disulfide-based cationic dextran conjugate having dextran as the main chain and disulfide-linked 1,4-bis(3-aminopropyl)piperazine (BAP) residues as the grafts was designed and successfully prepared as a multifunctional gene delivery vector for targeted gene delivery to ovarian cancer SKOV-3 cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service