Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

237264

Sigma-Aldrich

Resolve-Al La

99%

Synonym(s):

La(tmhd)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La(OCC(CH3)3CHCOC(CH3)3)3
CAS Number:
Molecular Weight:
688.71
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
370 °C (lit.)

Assay

99%

form

powder or crystals

bp

370 °C (lit.)

mp

128-131 °C (lit.)

SMILES string

CC(C)(C)C(=O)\C=C(/O[Yb](O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/3C11H20O2.La/c3*1-10(2,3)8(12)7-9(13)11(4,5)6;/h3*7,12H,1-6H3;/q;;;+3/p-3/b3*8-7-;

InChI key

IXHLTRHDDMSNAP-LWTKGLMZSA-K

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and electrical properties of a La2O3 thin film as a gate dielectric.
Jun JH, et al.
The Journal of the Korean Physical Society, 11(6) (2002)
MOCVD of lanthanum oxides from La (tmhd)3 and La (tmod)3 precursors: A thermal and kinetic investigation.
Bedoya C, et al.
Chem. Vap. Deposition, 12(1), 46-53 (2006)
Study on the precursors for La2O3 thin films deposited on silicon substrate.
Jun J, et al.
Journal of Materials Science Letters, 21(23), 1847-1849 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service