Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

232386

Sigma-Aldrich

(+)-2-Carene

97%

Synonym(s):

(1S)-3,7,7-Trimethylbicyclo[4.1.0]hept-2-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2038651
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]20/D +90.0°, c = 6 in ethanol

refractive index

n20/D 1.476 (lit.)

bp

167-168 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

SMILES string

CC1=C[C@H]2[C@@H](CC1)C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-9(6-7)10(8,2)3/h6,8-9H,4-5H2,1-3H3/t8-,9+/m1/s1

InChI key

IBVJWOMJGCHRRW-BDAKNGLRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-2-Carene is a natural bicyclic monoterpene that is commonly found in the essential oils of various plants, such as rosemary, cedarwood, and pine. In organic synthesis, it is used as a chiral building block for the synthesis of a variety of chiral compounds.

Application

(+)-2-Carene can be used as a chiral building block in the synthesis of chiral non-racemic 2,2-dimethyl-1,3-disubstituted cyclopropane derivatives. It is also used as a reactant in the enantio-selective synthesis of (+)-α-elemene.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Codruta Ignea et al.
Nature communications, 10(1), 3799-3799 (2019-08-25)
Synthetic biology efforts for the production of valuable chemicals are frequently hindered by the structure and regulation of the native metabolic pathways of the chassis. This is particularly evident in the case of monoterpenoid production in Saccharomyces cerevisiae, where the
Haocheng Liu et al.
Foods (Basel, Switzerland), 9(1) (2020-01-16)
Mangoes (Mangifera indica L.) are wildly cultivated in China with different commercial varieties; however, characterization of their aromatic profiles is limited. To better understand the aromatic compounds in different mango fruits, the characteristic aromatic components of five Chinese mango varieties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service