Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21987

Sigma-Aldrich

(1S,3R)-cis-4-Carene

≥95.0% (sum of enantiomers, GC)

Synonym(s):

(1R,4R)-4,7,7-Trimethylbicyclo[4.1.0]hept-2-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
3044038
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −145±5°, neat

refractive index

n20/D 1.469

density

0.849 g/mL at 20 °C (lit.)

SMILES string

[H][C@]12C[C@@H](C)C=C[C@@]1([H])C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-9(6-7)10(8,2)3/h4-5,7-9H,6H2,1-3H3/t7-,8+,9-/m0/s1

InChI key

LGNSZMLHOYDATP-YIZRAAEISA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service