MilliporeSigma
  • Using mass spectrometry to highlight structures of degradation compounds obtained by photolysis of chloroacetamides: case of acetochlor.
Using mass spectrometry to highlight structures of degradation compounds obtained by photolysis of chloroacetamides: case of acetochlor.
Journal of chromatography. A (2013-09-10)
Yasmine Souissi, Sophie Bourcier, Sélim Ait-Aissa, Emmanuelle Maillot-Maréchal, Stéphane Bouchonnet, Christophe Genty, Michel Sablier
ABSTRACT
The photooxidation of acetochlor (a pesticide belonging to the acetamides group) using a polychromatic UV irradiation in ultrapure water was studied. This study reports the efficiency of mass spectrometry for the characterization of photodegradation products of acetochlor. Decompositions of protonated ions MH+are proposed in electrospray (ESI) mode for LC–MS, while electron ionization (EI) and chemical ionization modes (CI) are used for GC–MS. The knowledge of fragmentation and the use of a combination of experiments (MS/MS, high resolution) allow the characterization of photoproducts. Structural elucidation is assisted by the use of photolysed deuterated compounds. Fifteen major degradation products have been characterized, five by LC-QTOF, six photoproducts by GC-ITMS, and four are observed by both techniques. In vitro bioassays based on the quantification of receptor-mediated activity demonstrated that acetochlor photolysis engenders a moderate but significant estrogenic activity. Moreover, a quantitative structure–activity relationship (QSAR) approach was used to assess the potential toxicity effect of acetochlor and its by-products. The predictions were analyzed showing a variety of toxicity profiles of acetochlor photoproducts depending on the toxicological investigated endpoint.
Sigma-Aldrich
2-Chloroacetamide, ≥98.0% (HPLC)
Supelco
Acetochlor, PESTANAL®, analytical standard
Sigma-Aldrich
2-Chloroacetamide, ≥98%