Skip to Content
MilliporeSigma
All Photos(2)

Documents

222682

Sigma-Aldrich

Bis(4-fluorophenyl)methanol

98%

Synonym(s):

4,4′-Difluorobenzhydrol, Bis(4-fluorophenyl) carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(FC6H4)2CHOH
CAS Number:
Molecular Weight:
220.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

143 °C/3 mmHg (lit.)

mp

43-45 °C (lit.)

SMILES string

OC(c1ccc(F)cc1)c2ccc(F)cc2

InChI

1S/C13H10F2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8,13,16H

InChI key

WCTZPQWLFWZYJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(4-fluorophenyl)methanol is one of the degardation product of flunarizine hydrochloride (FLZ).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dina T El-Sherbiny et al.
Journal of separation science, 28(2), 197-202 (2005-03-10)
The separation of flunarizine hydrochloride (FLZ) and five of its degradation products--1-[bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)piperazine, 4-oxide (A), bis(4-fluorophenyl)methanone (B), bis(4-fluorophenyl)methanol (C), 1-(3-phenyl-2-propenyl)piperazine(D), and 1-[bis-4-fluorophenyl) methyl] piperazine (E)--could be accomplished by reversed phase liquid chromatography using either micellar or microemulsion mobile phases. Cyanopropyl-bonded stationary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service