Skip to Content
MilliporeSigma
All Photos(1)

Documents

216151

Sigma-Aldrich

1-Methoxy-1,4-cyclohexadiene

technical grade, 85%

Synonym(s):

2,5-Dihydroanisole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O
CAS Number:
Molecular Weight:
110.15
Beilstein:
1925467
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

148-150 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

COC1=CCC=CC1

InChI

1S/C7H10O/c1-8-7-5-3-2-4-6-7/h2-3,6H,4-5H2,1H3

InChI key

RGXIXVNKYUAKHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methoxy-1,4-cyclohexadiene reacts with RuCl3·xH2O in an alcohol solvent to give [RuCl26-C6H5OR)2 (R = Me, Et or HOCH2CH2).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Convenient One-Pot Synthesis of a Functionalized-Arene Ruthenium Half-Sandwich Compound [RuCl2 (?6-C6H5OCH2CH2OH)] 2.
Soleimannejad J and White C.
Organometallics, 24(10), 2538-2541 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service