Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

206164

Sigma-Aldrich

Rhodium on carbon

greener alternative

extent of labeling: 5 wt. % loading

Synonym(s):

Rh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rh
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

core: rhodium
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

5 wt. % loading

greener alternative category

SMILES string

[Rh]

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Rhodium on carbon (Rh/C) can be used as a catalyst in the reductive transformation of carbonyl compounds. It can be used as a heterogeneous catalyst for the production of formaldehyde through a greener technique.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium supported on activated carbon as a heterogeneous catalyst for hydroformylation of propylene in supercritical carbon dioxide
Dharmidhikari S and Abraham MA
Journal of Supercritical Fluids, 18(1), 1-10 (2000)
Reductive Transformations of Carbonyl Compounds Catalyzed by Rhodium Supported on a Carbon Matrix by using Carbon Monoxide as a Deoxygenative Agent
Yagafarov NZ, et al.
ChemCatChem, 7(17), 2590-2593 (2015)
William J S Lockley et al.
Journal of labelled compounds & radiopharmaceuticals, 63(13), 531-552 (2020-09-05)
Catalytic dehalogenation of aromatic halides using isotopic hydrogen gas is an important strategy for labelling pharmaceuticals, biochemicals, environmental agents and so forth. To extend, improve and further understand this process, studies have been carried out on the scrambling of deuterium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service