Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

205370

Sigma-Aldrich

Dimethyl aminoterephthalate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CO2CH3)2
CAS Number:
Molecular Weight:
209.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

127-130 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccc(c(N)c1)C(=O)OC

InChI

1S/C10H11NO4/c1-14-9(12)6-3-4-7(8(11)5-6)10(13)15-2/h3-5H,11H2,1-2H3

InChI key

DSSKDXUDARIMTR-UHFFFAOYSA-N

General description

The tricarbonylchromium complex of dimethyl aminoterephthalate was prepared and studied.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahaffy CA.
Synth. React. Inorg. Met.-Org. Chem. , 14(6), 895-903 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service