Skip to Content
MilliporeSigma
All Photos(2)

Documents

203300

Sigma-Aldrich

Gadolinium(III) sulfate octahydrate

≥99.99% trace metals basis

Synonym(s):

Gadolinium(3+) trisulfate octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd2(SO4)3 · 8H2O
CAS Number:
Molecular Weight:
746.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: gadolinium

impurities

≤100.0 ppm Trace Rare Earth Analysis

density

3.01 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.[Gd+3].[Gd+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Gd.3H2O4S.8H2O/c;;3*1-5(2,3)4;;;;;;;;/h;;3*(H2,1,2,3,4);8*1H2/q2*+3;;;;;;;;;;;/p-6

InChI key

JBFCVGZGWFIPNK-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service