Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197688

Sigma-Aldrich

3-(3,4-Dimethoxyphenyl)-1-propanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3(CH2)3OH
CAS Number:
Molecular Weight:
196.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

refractive index

n20/D 1.54 (lit.)

bp

142-144 °C/0.5 mmHg (lit.)

density

1.081 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

COc1ccc(CCCO)cc1OC

InChI

1S/C11H16O3/c1-13-10-6-5-9(4-3-7-12)8-11(10)14-2/h5-6,8,12H,3-4,7H2,1-2H3

InChI key

ZISWRXJZUKDIOO-UHFFFAOYSA-N

Application

3-(3,4-Dimethoxyphenyl)-1-propanol was used in the synthesis of fluorinated benzamide neuroleptic, (S)-N-[(1-ethyl-2- pyrrolidinyl)methyl]-5-(3-fluoropropyl)-2,3-dimethoxybenzamide[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Mukherjee
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes, 42(8), 713-721 (1991-01-01)
Synthesis of a fluorinated benzamide neuroleptic, (S)-N-[(1-ethyl-2- pyrrolidinyl)methyl]-5-(3-fluoropropyl)-2,3- dimethoxybenzamide starting from 3-(3,4-dimethoxyphenyl)-1-propanol in 20-25% overall yield is reported. Radiosynthesis of (S)-N-[(1-ethyl-2-pyrrolidinyl)methyl]- 5-(3[18F]fluoropropyl)-2-methoxybenzamide([18F]FPHB) and (S)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(3[18F]fluoropropyl)-2,3- dimethoxybenzamide([18F]FPHB) was carried out by nucleophilic substitution reaction of (S)-N-[(1-ethyl-2-pyrrolidinyl)methyl-5-(3-tosyloxypropyl)-2-me tho xybenzamide and (S)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(3-tosyloxypropyl)-2,3- dimethoxybenzamide respectively

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service