Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

142328

Sigma-Aldrich

3-(4-Methoxyphenyl)-1-propanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4(CH2)3OH
CAS Number:
Molecular Weight:
166.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.532 (lit.)

bp

164-168 °C/18 mmHg (lit.)

mp

26 °C (lit.)

functional group

hydroxyl

SMILES string

COc1ccc(CCCO)cc1

InChI

1S/C10H14O2/c1-12-10-6-4-9(5-7-10)3-2-8-11/h4-7,11H,2-3,8H2,1H3

InChI key

NIIDHUCLROLCBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(4-Methoxyphenyl)-1-propanol participates in β-alkylation of 1-phenylethanol catalyzed by RuCl2(DMSO)4.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

RuCl2(DMSO)4 catalyzes the ?-alkylation of secondary alcohols with primary alcohols through a hydrogen autotransfer process.
Martinez R, et al.
Tetrahedron, 62(38), 8982-8987 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service