Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

192759

Sigma-Aldrich

Ethylaluminum dichloride solution

25 wt. % in toluene

Synonym(s):

Dichloroethylaluminum, Dichloromonoethylaluminum, Ethylaluminium dichloride, Ethylaluminum dichloride (6CI), Ethyldichloroaluminium, Ethyldichloroaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2AlCl2
CAS Number:
Molecular Weight:
126.95
Beilstein:
4123357
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5 mmHg ( 60 °C)

form

liquid

concentration

25 wt. % in toluene

density

0.927 g/mL at 25 °C

SMILES string

CC[Al](Cl)Cl

InChI

1S/C2H5.Al.2ClH/c1-2;;;/h1H2,2H3;;2*1H/q;+2;;/p-2

InChI key

UAIZDWNSWGTKFZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup - Solvent

Flash Point(C)

4 °C - closed cup - Solvent


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Barry B Snider et al.
The Journal of organic chemistry, 68(16), 6451-6454 (2003-08-05)
EtAlCl(2)-induced cyclization of chiral gamma,delta-unsaturated ketones 11c and 17b takes place mainly from the expected face. The selectivity is modest for 11c (60:40) in which the large substituent is a primary alkyl group and the medium substituent is a methyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service