Skip to Content
MilliporeSigma
All Photos(2)

Documents

182532

Sigma-Aldrich

Poly(vinylbenzyl chloride), 60/40 mixture of 3- and 4-isomers

average Mn ~55,000, average Mw ~100,000 by GPC/MALLS, powder

Synonym(s):

PVBC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4CH2Cl)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn ~55,000
average Mw ~100,000 by GPC/MALLS

SMILES string

ClCc1ccc(C=C)cc1.ClCc2cccc(C=C)c2

InChI

1S/C27H29Cl3/c1-20(25-11-7-23(18-29)8-12-25)16-27(26-13-9-24(19-30)10-14-26)15-6-21-2-4-22(17-28)5-3-21/h2-5,7-14,20,27H,6,15-19H2,1H3

InChI key

QBVHDIKGDQTDMC-UHFFFAOYSA-N

Application

Poly(vinyl benzyl chloride) can be used:
  • As a precursor to fabricate alkaline anion exchange membrane water electrolyzers(AAEMWE).
  • As a hydroxide conducting polymer to prepare anion exchange membranesfor fuel cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service