Skip to Content
MilliporeSigma
All Photos(2)

Documents

181153

Sigma-Aldrich

Nylon 11

pellets, 3 mm

Synonym(s):

Polyundecanolactam

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-NH(CH2)10CO-]n
CAS Number:
Molecular Weight:
201.31
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

particle size

3 mm

transition temp

Tm 198 °C
Tg 46 °C

density

1.026 g/mL at 25 °C (lit.)

SMILES string

NCCCCCCCCCCC(O)=O

InChI

1S/C33H65N3O3/c34-28-22-16-10-4-2-8-14-20-26-32(38)36-30-24-18-12-6-3-9-15-21-27-33(39)35-29-23-17-11-5-1-7-13-19-25-31-37/h31H,1-30,34H2,(H,35,39)(H,36,38)

InChI key

IAPYCAVXWCDYGR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Valeria Bugatti et al.
Polymers, 11(4) (2019-04-10)
Antimicrobial packaging systems are recognized as effective approaches to prolong food shelf life. In this context, Bio-based PA11 loaded with a food-grade zeolite were prepared using ball milling technology in the dry state. Zeolite was filled with sodium salicylate, as
Valeria Bugatti et al.
Nanomaterials (Basel, Switzerland), 8(3) (2018-03-02)
Bio-based membranes were obtained using Polyamide 11 (PA11) from renewable sources and a nano-hybrid composed of halloysite nanotubes (HNTs) filled with lysozyme (50 wt % of lysozyme), as a natural antimicrobial molecule. Composites were prepared using an electrospinning process, varying
María Elena Sánchez-Vergara et al.
Polymers, 12(1) (2019-12-22)
Chemical degradation is a major disadvantage in the development of organic semiconductors. This work proposes the manufacture and characterization of organic semiconductor membranes in order to prevent semiconductor properties decreasing. Semiconductor membranes consisting of Nylon-11 and particles of π-conjugated molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service