Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

180238

Sigma-Aldrich

Borane dimethylamine complex

97%

Synonym(s):

Dimethylamine borane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH · BH3
CAS Number:
Molecular Weight:
58.92
EC Number:
MDL number:
UNSPSC Code:
12352402
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: reductant

mp

36 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

[BH3-][NH+](C)C

InChI

1S/C2H10BN/c1-4(2)3/h4H,1-3H3

InChI key

RUOMFVPJBOADHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Borane dimethylamine complex can be used as a reducing agent in various applications. Examples are:
  • Ni-catalyzed reduction of electron-deficient aryl tosylates.
  • Pd-catalyzed reduction of nitroarenes to corresponding amines.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Eckardt et al.
ChemSusChem (2020-04-22)
The influence of different additives on the oxygen reduction reaction/oxygen evolution reaction (ORR/OER) in magnesium-containing N-butyl-N-methylpyrrolidinium bis(trifluoromethanesulfonyl)imide ([BMP][TFSI]) on a glassy carbon electrode was investigated to gain a better understanding of the electrochemical processes in Mg-air batteries. 18-Crown-6 was used
Tetrahedron Letters, 47, 7515-7515 (2006)
Daniel G W Alanine et al.
Cell, 178(1), 216-228 (2019-06-18)
The Plasmodium falciparum reticulocyte-binding protein homolog 5 (PfRH5) is the leading target for next-generation vaccines against the disease-causing blood-stage of malaria. However, little is known about how human antibodies confer functional immunity against this antigen. We isolated a panel of
Panagiota Kalomoiri et al.
Chembiochem : a European journal of chemical biology, 21(23), 3338-3348 (2020-07-16)
The controlled self-assembly of peptide- and protein-based pharmaceuticals is of central importance for their mode of action and tuning of their properties. Peptide YY3-36 (PYY3-36 ) is a 36-residue peptide hormone that reduces food intake when peripherally administered. Herein, we
Junlong Tian et al.
Scientific reports, 5, 8025-8025 (2015-01-27)
A carbon-matrix nickel composite magnetoplasmonic film with a 3D sub-micron periodic triangular roof-type antireflection structure (SPTAS) was fabricated via a simple and promising method that combines chemosynthesis with biomimetic techniques. The Troides helena (Linnaeus) forewing (T_FW) was chosen as the

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service