Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

178985

Sigma-Aldrich

Borane trimethylamine complex

97%

Synonym(s):

NSC 10220, NSC 145941, NSC 53323, Trihydro(trimethylamine)boron, Trimethylamine borane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N · BH3
CAS Number:
Molecular Weight:
72.95
Beilstein:
3905060
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: reductant

bp

172 °C (lit.)

mp

92-94 °C (lit.)

functional group

amine

SMILES string

[BH3-][N+](C)(C)C

InChI

1S/C3H12BN/c1-5(2,3)4/h1-4H3

InChI key

KQVSXUGMDCAISO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reducing agent for the preparation of plate-type palladium-zinc oxide catalyst

Reagent for chemical vapor generation and spectrometry determination of elements using amineboranes and cyanotrihydroborate(III) reagents

Catalyst for:
  • Regioselective reductive opening of benzylidene acetal glycosides
  • Polarity reversal

Used to study
  • Electrodeposition condition effects on magnetism of cobalt-iron magnetic films
  • Boron-nitrogen bond dissociation energies and physiological activities of boronated compounds
Stable, easy-to-handle source of borane for the reduction of ketones and Schiff bases and for the hydroboration of olefins.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 59, 705-705 (1994)
Analytical Chemistry, 66, 4283-4283 (1994)
Aldrichimica Acta, 6, 51-51 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service